Imprimer
THELON XXG GTX S3 HI CI ESD Safety Shoes, Lace-Up, Low
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
728611/39 728611/40 728611/41 728611/42 728611/43 728611/44 728611/45 728611/46 728611/47 728611/48
ELTN728611/39EA 127.9 EUR
ELTN728611/39 ELTN728611/40 ELTN728611/41 ELTN728611/42 ELTN728611/43 ELTN728611/44 ELTN728611/45 ELTN728611/46 ELTN728611/47 ELTN728611/48
THELON XXG GTX S3 HI CI ESD Safety Shoes, Lace-Up, Low
Chaussures et bottes
Safety shoe, THELON XXG GTX, in brown finish.
  • Hydrophobized nubuck leather
  • GORE-TEX performance comfort footwear
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Non-metallic, penetration-resistant midsole
  • Rubber/PU sole WELLMAXX GRIP
  • Composite toe cap
For indoors and outdoor use, in areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), or areas where there is a risk of electrostatic discharge (ESDS/ESD). For workplaces on hard undergrounds, the revolutionary Infinergy® sole core cushions impacts and provides a rebound effect when the compressive impulse subsides, resulting in more energy in every step.
Certifications: EN ISO 20345 S3 CI/HI/HRO/SRC, form A.