Imprimer
STEWART XXG Pro GTX S2 HI CI ESD Safety Boots, Lace-Up, Mid
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
768671/39 768671/40 768671/41 768671/42 768671/43 768671/44 768671/45 768671/46 768671/47 768671/48
ELTN768671/39EA 142.9 EUR
ELTN768671/39 ELTN768671/40 ELTN768671/41 ELTN768671/42 ELTN768671/43 ELTN768671/44 ELTN768671/45 ELTN768671/46 ELTN768671/47 ELTN768671/48
STEWART XXG Pro GTX S2 HI CI ESD Safety Boots, Lace-Up, Mid
Chaussures et bottes
Safety boot, STEWART XXG Pro GTX, in black/red finish.
  • Hydrophobized nubuck leather
  • GORE-TEX performance comfort footwear
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Non-metallic, penetration-resistant midsole
  • Rubber/PU sole WELLMAXX GRIP
  • TPU scuff cap
  • Composite toe cap
For indoor and outdoor use, in areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), or areas where there is a risk of electrostatic discharge (ESDS/ESD). For workplaces on hard undergrounds, the revolutionary Infinergy® sole core cushions impacts and provides a rebound effect when the compressive impulse subsides, resulting in more energy in every step.
Certifications: EN ISO 20345 S3 CI/HI/HRO, form B.