Imprimer
SHARKI S1 ESD Safety Shoes, Lace-up, Low
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
720840/36 720840/37 720840/38 720840/39 720840/40 720840/41 720840/42 720840/43 720840/44 720840/45 720840/46 720840/47 720840/48 720845/36 720845/37 720845/38 720845/39 720845/40 720845/41 720845/42 720845/43 720845/44 720845/45 720845/46 720845/47 720845/48
ELTN720840/36EA 87.9 EUR
ELTN720840/36 ELTN720840/37 ELTN720840/38 ELTN720840/39 ELTN720840/40 ELTN720840/41 ELTN720840/42 ELTN720840/43 ELTN720840/44 ELTN720840/45 ELTN720840/46 ELTN720840/47 ELTN720840/48 ELTN720845/36 ELTN720845/37 ELTN720845/38 ELTN720845/39 ELTN720845/40 ELTN720845/41 ELTN720845/42 ELTN720845/43 ELTN720845/44 ELTN720845/45 ELTN720845/46 ELTN720845/47 ELTN720845/48
SHARKI S1 ESD Safety Shoes, Lace-up, Low
Chaussures et bottes
ESD safety shoe, SHARKI, available in blue or red finish. Low, lace-up S1.
  • Microfibre material
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole ESD
  • Soft fleece ESD insole
  • Rubber/TPU/EVA sole TRANSFOAMERS
  • Composite toe cap
For dry work areas, applications include industry, storage, transport, assembly etc. (S1). Also for areas where there is a risk of electrostatic discharge (ESDS/ESD).
Certifications: EN ISO 20345 S1 HRO/SRC, form A.