Imprimer
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
OmniTop Sample Tubes®
PAWBEVSS-PVC7.5MLEA 1350 EUR
PAWBEVSS-PVC7.5ML PAWBEVSS-CFLEX6.75 PAWBEVSS-PVC4.24 PAWBEVSS-CFLEX5.25 PAWBEVSS-PVC5.74 734-3608 PAWBEVSS-PVC2.0ML 734-3609 734-3457 PAWBEVSS-PVC10.5 734-3611 734-3458 734-3612 734-3459 734-3613 734-3614 734-3453 PAWBEVSS-CFLEX3.0 734-3454 PAWBEVSS-PVC5.5ML 734-3455 734-3610 734-3456 PAWBEVSS-PVC10.75 PAWBEVSS-CFLEX7.0 734-3450 PAWBEVSS-PVC9.5ML 734-3451 PAWBEVSS-CFLEX3.5 734-3452 PAWBEVSS-CFLEX8.25 PAWBEVSS-PVC9.0ML PAWBEVSS-PVC2.74 PAWBEVSS-CFLEX8.5 PAWBEVSS-CFLEX1.75 PAWBEVSS-PVC10.0 PAWBEVSS-PVC1.24 PAWBEVSS-PVC9.24 PAWBEVSS-PVC3.5ML 734-3589 734-3468 734-3469 734-3585 734-3464 734-3586 734-3465 734-3466 734-3587 PAWBEVSS-CFLEX9.75 PAWBEVSS-PVC7.0ML 734-3588 734-3467 734-3460 734-3461 734-3462 734-3463 734-3590 734-3591 734-3470 PAWBEVSS-CFLEX8.75 PAWBEVSS-CFLEX7.25 PAWBEVSS-CFLEX0.75 PAWBEVSS-PVC6.0ML PAWBEVSS-PVC2.24 PAWBEVSS-PVC3.74 734-3479 PAWBEVSS-PVC1.5ML 734-3596 734-3475 734-3476 734-3597 734-3598 734-3477 PAWBEVSS-CFLEX0.5 PAWBEVSS-CFLEX4.0 734-3478 734-3599 734-3592 734-3471 734-3472 734-3593 734-3594 734-3473 PAWBEVSS-CFLEX4.5 734-3474 734-3595 PAWBEVSS-CFLEX8.0 PAWBEVSS-CFLEX5.5 734-3480 734-3481 PAWBEVSS-CFLEX9.5 PAWBEVSS-CFLEX3.75 PAWBEVSS-CFLEX2.25 PAWBEVSS-PVC8.74 PAWBEVSSCFLEX10.24 PAWBEVSS-PVC4.0ML PAWBEVSS-PVC0.74 PAWBEVSS-PVC7.24 734-3486 734-3487 734-3488 734-3489 734-3482 734-3483 734-3484 734-3485 PAWBEVSS-CFLEX9.25 PAWBEVSS-PVC1.74 734-3490 734-3491 734-3492 PAWBEVSS-CFLEX2.75 PAWBEVSS-PVC6.5ML PAWBEVSS-CFLEX1.25 PAWBEVSS-CFLEX10.1 PAWBEVSS-PVC8.24 PAWBEVSS-PVC8.5ML PAWBEVSS-PVC9.74 PAWBEVSS-CFLEX1.0 734-3497 PAWBEVSS-PVC1.0ML 734-3498 734-3499 PAWBEVSS-CFLEX5.0 734-3493 734-3494 PAWBEVSS-CFLEX1.5 734-3495 PAWBEVSS-CFLEX9.0 734-3496 PAWBEVSS-CFLEX6.5 PAWBEVSS-PVC5.24 PAWBEVSS-CFLEX5.75 PAWBEVSS-PVC4.5ML PAWBEVSS-PVC6.74 PAWBEVSS-CFLEX4.25 PAWBEVSS-CFLEX10.4 PAWBEVSS-PVC8.0ML PAWBEVSS-PVC10.25 PAWBEVSS-PVC2.5ML PAWBEVSS-CFLEX4.75 PAWBEVSS-PVC7.74 PAWBEVSS-CFLEX3.25 PAWBEVSS-PVC6.24 PAWBEVSS-CFLEX11.1 PAWBEVSS-PVC11.1 PAWBEVSS-CFLEX2.0 PAWBEVSS-CFLEX2.5 PAWBEVSS-CFLEX6.0 PAWBEVSS-PVC5.0ML PAWBEVSS-CFLEX7.75 PAWBEVSS-PVC3.24 PAWBEVSS-CFLEX7.5 PAWBEVSS-CFLEX6.25 PAWBEVSS-PVC0.5ML PAWBEVSS-PVC4.74 734-3604 734-3605 734-3606 PAWBEVSSCFLEX10.74 734-3607 734-3446 734-3600 734-3601 734-3447 734-3448 734-3602 734-3603 734-3449 734-3444 734-3445 PAWBEVSS-PVC3.0ML
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Tubes Tubes à essais
Optimise sampling processes with the Exact Volume Sampling Systems (EVSS) from OmniTop Sample Tubes®.
  • Customised sample volume at point of use
  • Aseptic, closed system sampling reduces contamination risk
  • Improved product yield
  • Reduced assembly and installation time
  • Easy operation and closed system transfer
Choose any volume between 0,5 to 11,0 ml and take exact volume samples with a ±0,25 ml accuracy. The EVSS is available with a C-Flex® tubing inlet or a PVC (Tygon®) tubing option with a male Luer end connection.
The EVSS can be aseptically connected to a bioreactor or other device using standard tube welders. It can also be aseptically sealed and removed (wet or dry) using tubing sealers such as the Genesis SE340 or handheld battery powered version SE640.
Certifications: Sterility Assurance Level (SAL) 10⁻⁶ for single-use assemblies. All accessories are Animal Derived Component Free (ADCF).
Informations sur les accessoires: The single-use assemblies are individually double bagged and gamma Irradiated to Sterility Assurance Level (SAL) 10⁻⁶. The sampling rack is reusable and can be found under the accessories tab below.
Informations de commande: The sampling rack is not included with the single-use assemblies and must be ordered separately.