Imprimer
Jenway® Dissolved Oxygen Probes
Fournisseur: Cole-Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
99964-71 99964-91
JENW99964-71EA 574.12 EUR
JENW99964-71 JENW99964-91
Jenway® Dissolved Oxygen Probes
Électrodes Sondes à oxygène dissous
Jenway has an extensive range of dissolved oxygen probes suitable for portable or bench dissolved oxygen meters.
  • The probes for the dissolved oxygen meters have all been updated in order to unify the range
The dissolved oxygen probes have been redesigned so that they are all the same diameter. As a result of this redesign, the new probes have a narrower diameter than the old probes. The new probes are easily identified with a B after the probe part number on the probe or meter packaging.