Imprimer
Cole-Parmer® BB-200 Electric Bunsen Burners
Fournisseur: Cole-Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
36135-02 36135-01
ELET36135-02EA 1420 EUR
ELET36135-02 ELET36135-01
Cole-Parmer® BB-200 Electric Bunsen Burners
Appareils chauffants Brûleurs à gaz
Provide clean and easy operation.
  • Top cowl deflects heat away from hands
  • Air circulation from vented housing keeps base cool to touch
  • Energy efficient - only consume 400 W of power
These electric Bunsen burners combine the advantages of a regular gas burner with the clean and easy operation of a heating mantle. Corrosion-resistant stainless steel burners features a cone-shaped coiled resistance heater which directs radiant heat out from the cavity. Cooling vents circulate air to cool the base, making it possible to hold while in use. A detachable cowl also deflects heat from the medium surface.
Ideal for noncontact radiant heating of materials in test tubes, crucibles, small flasks, and beakers. Effective heating of a test tube sample should be done 25 mm above the top of the burner. Crucible heating may be achieved by removing the cowl.
Note: To regulate the power input and temperature of the Bunsen burner, a heating controller is required. without a controller, the burner operates continuously at full heating power. The Bunsen burner can be purchased with or without the recommended controller.