Imprimer
Cleanroom Trousers, Movix Anti-static
Fournisseur: ALSICO HIGHTECH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1033614-XS 1033614-S 1033614-M 1033614-L 1033614-XL 1033614-XXL 1033614-3XL
ALSO1033614-XSEA 48.3 EUR
ALSO1033614-XS ALSO1033614-S ALSO1033614-M ALSO1033614-L ALSO1033614-XL ALSO1033614-XXL ALSO1033614-3XL
Cleanroom Trousers, Movix Anti-static
Tenue de travail
These trousers are made of Movix (98% polyester, 2% carbon). Suitable for C and D classes or ISO 7 to 9 cleanrooms. For higher cleanroom classes worn as an undergarment piece under a coverall. Can also be used for clean areas.
  • Elasticated waistband with drawstring
  • Side pockets
  • Cuffs in main fabric
Certifications: IEST-RP-CC003.5.