Imprimer
Cryo.s Cryogenic Storage Tubes with Datamatrix
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
122263-2DG_500 123263-2DG_500 126263-2DG_500 127263-2DG_300
479-2440EA 944 EUR
479-2440 479-2442 479-2444 479-2445
Cryo.s Cryogenic Storage Tubes with Datamatrix
Fioles
Manufactured from high quality polypropylene, Cryo.s freezing tubes are suitable for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin such as blood, serum, and sperm, as well as antibodies, RNA, DNA, and protein samples.
  • Sterile
  • Endotoxin-free
  • Free of RNase, DNase, and human DNA
  • Non-pyrogenic and non-cytotoxic
These cryogenic sample tubes are manufactured from high quality polypropylene, which means they are ideally suited for use over a broad range of temperatures (–196 to 121 °C).
Laser etched, ECC200 code is the newest version of Datamatrix coding that is available on Greiner Bio One tubes. Datamatrix coding supports advanced error correction algorithms with capability to decipher partially damaged codes.
Compatible with automated liquid handling and de-/re-capping systems.
Attention: Cryo.s tubes are not intended for sample storage within reproductive medicine.