Imprimer
Cole Parmer® SC-200 Sample Concentrators
Fournisseur: Cole-Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
36620-42 36620-43 36620-97 36620-98 36620-99 36620-67 99950-07
TECN36620-42EA 1610 EUR
TECN36620-42 TECN36620-43 TECN36620-97 TECN36620-98 TECN36620-99 TECN36620-67 TECN99950-07
Cole Parmer® SC-200 Sample Concentrators
Chauffe-blocs Concentrateurs d'échantillon
Quickly concentrate multiple samples in a block at once.
  • Variable needle position suits various Dri-Block® inserts
  • Small size makes it ideal for use under a fume hood
  • Needles, block, and block heater sold separately
Concentrator blows an inert gas over the surface of your samples via stainless steel needles (not included). Blown gas in combination with the heat from the heater produce ideal conditions for fast efficient evaporation. Insert needles into a silicone membrane in any configuration. Fully adjustable gas reservoir provides accurate height control. Maximum vertical traveling distance is 32 cm. Maximum gas usage is 15 L/min.
Informations de commande: Needles, block, and block heater sold separately. Order PTFE-coated needles for use with corrosive samples.
Informations de livraison: Supplied with gas reservoir and stand.