Imprimer
RNA ScreenTape Analysis
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5067-5576 5067-5577 5067-5578
732-3997EA 210 EUR
732-3997 732-3998 732-3999
RNA ScreenTape Analysis
Systèmes d'électrophorèse Capillaires pour électrophorèse
Reliable, efficient, and objective measurements of quality and quantity of RNA samples.
  • Utilises RINe - a user independent algorithm for accurate and objective assessment of eukaryotic or prokaryotic total RNA integrity
  • Provides fast results with data delivered in approximately 1 minute per sample
  • Minimal sample consumption is required as only 1 µl of sample is needed for accurate data
  • RNA sample detection down to 5 ng/µl is possible
  • Objective quality control of precious RNA samples is enabled within RNAseq, microarray analysis, and quantitative RT-PCR workflows
  • Ready-to-use consumables provide excellent reproducibility
  • No cross contamination and sample carryover by using single lanes and individual loading tips
  • Partially used ScreenTape devices can be used for completion at a later point in time
RNA ScreenTape analysis provides efficient and reliable electrophoretic separation of total RNA down to 5 ng/µl. Compatible with eukaryotic and prokaryotic samples, the assay provides accurate information about sample integrity and quantity. Objective evaluation of RNA degradation is delivered with an RNA integrity number (RINe) that can be used for sample quality control in NGS workflows.
The assay is compatible with all Agilent TapeStation instruments.
Note: Some products within this assay contain SYBR™ Gold, which is licensed from Life Technologies Corporation for use in research and development only. SYBR™ is a registered trademark of Life Technologies Corporation.
Attention: For research use only. Not for use in diagnostic procedures.