Imprimer
High Sensitivity DNA ScreenTape Analysis (ScreenTape and Reagents)
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5067-5584 5067-5585 5067-5587 5067-5592 5067-5593 5067-5594 5067-5603
AGLS5067-5584EA 358 EUR
AGLS5067-5584 AGLS5067-5585 AGLS5067-5587 AGLS5067-5592 AGLS5067-5593 AGLS5067-5594 AGLS5067-5603
High Sensitivity DNA ScreenTape Analysis (ScreenTape and Reagents)
Systèmes d'électrophorèse Capillaires pour électrophorèse
DNA fragment or library analysis from samples of limited abundance, for samples up to 5000 bp.
  • The broad size range allows analysis of DNA fragments or libraries from 35 to 5000 bp
  • Results are obtained quickly, in approximately 1 minute per sample
  • Detection of DNA samples with limited abundance is possible, for samples down to 5 pg/µl
  • Ready-to use ScreenTape devices, pre-packaged reagent and automated data analysis offer an easy to use system with reproducible DNA sizing and quantification results
  • Reliable sample quality control is provided for next-generation sequencing (NGS) workflows
  • No cross contamination or sample carryover is caused due to the use of single lanes and individual loading tips
  • Partially used ScreenTape devices can be used to completion at a later point in time
Agilent's TapeStation High Sensitivity DNA system provides a fast, automated, and flexible solution for samples of limited abundance in many applications. High Sensitivity DNA electrophoresis can be used for quality control of next-generation sequencing (NGS) libraries.
The High Sensitivity DNA ScreenTape assays are suitable for analysis of precious samples. They can separate DNA fragments and libraries up to 5000 base pairs in 1 to 2 minutes per sample. Data output yields objective quantitative and purity measurements from only 2 µl of sample.
Assays are compatible with all Agilent TapeStation instruments.
Note: Some products within these assays contain SYBR™ Green I, which is licensed from Life Technologies Corporation for use in research and development only. SYBR™ is a registered trademark of Life Technologies Corporation.
Attention: For research use only. Not for use in diagnostic procedures.