Imprimer
Small Fragment and NGS Kits
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DNF-473-0500 DNF-473-1000 DNF-474-0500 DNF-474-1000 DNF-476-0500 DNF-477-0500
AGLSDNF-473-0500EA 1470 EUR
AGLSDNF-473-0500 AGLSDNF-473-1000 AGLSDNF-474-0500 AGLSDNF-474-1000 AGLSDNF-476-0500 AGLSDNF-477-0500
Small Fragment and NGS Kits
Systèmes d'électrophorèse Capillaires pour électrophorèse
Quality control for DNA smears and fragments including cfDNA analysis and NGS library QC.
  • Smear concentration range - The HS NGS and HS small fragment kits have a smear concentration range of 50 to 5000 pg/µl, The NGS and small fragment kits covers a higher range of 5 to 100 ng/µl
  • Fragment concentration range - The HS NGS and HS small fragment kits have a fragment concentration range of 5 to 500 pg/µl, The NGS and small fragment kits covers a higher input range of 0,1 to 10 ng/µl
  • Reliable sizing - kits for small and standard sized DNA fragments and smears help ensure accurate and precise sizing
  • Specific sizing ranges - The small fragment kits have a sizing range of 50 to 1500 bp, while the NGS kits cover 100 to 6000 bp, ensuring accurate sizing for specific applications
  • Low sample volume - Only 2 µl of sample is required, minimising sample loss to QC steps
The NGS and small fragment kits simplify NGS library quality control and analysis of DNA smears and fragments. These kits provide accurate quantification and sizing, saving you time during NGS workflows.
The small fragment kits cover a sizing range of 50 to 1500 bp, making them ideal for the analysis of small fragments, cfDNA, and small RNA library sample QC. The NGS fragment kits have a sizing range that is extended to 6000 bp and focuses on NGS library quality control. The small fragment and NGS fragment kits are specific for high input concentration ranges, while the HS small fragment and HS NGS fragment kits are designed for a lower concentration range.
Attention: For research use only. Not for use in diagnostic procedures.