Imprimer
VWR®, Pasteur Pipettes
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
612-7173EA 95.4 EUR
612-7173 612-7174
VWR®, Pasteur Pipettes
Pipettes Pipettes Pasteur
Disposable Pasteur pipettes made from AR® glass.
  • Non sterile
  • Automated manufacturing process ensures total production uniformity
  • Tapered upper opening for cotton ball plugging
Jet inner diameter: ~1 mm, body length: 90 mm, body diameter: 7 mm.
Certifications: Produced according to ISO 7712.