Imprimer
Masterflex® Nylon Clamps for Sanitary Fittings, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX00424-TNEA 293 EUR
MFLX00424-TN MFLX00424-TO MFLX12805-05
Masterflex® Nylon Clamps for Sanitary Fittings, Avantor®
Pinces Clips pour tubes
Collared clamp helps prevent unintentional opening.
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics