Imprimer
Value Plastics® RQ Series Quick Disconnect Fittings
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40618-48BLK 40618-49BLK 40618-50BLK 40618-51BLK 40618-52BLK 40618-53BLK 40618-55BLK 40618-56BLK 40618-44BLK 40618-45BLK 40618-46BLK 40618-47BLK
MFLX40618-48BLKEA 3260 EUR
MFLX40618-48BLK MFLX40618-49BLK MFLX40618-50BLK MFLX40618-51BLK MFLX40618-52BLK MFLX40618-53BLK MFLX40618-55BLK MFLX40618-56BLK MFLX40618-44BLK MFLX40618-45BLK MFLX40618-46BLK MFLX40618-47BLK
Value Plastics® RQ Series Quick Disconnect Fittings
Raccords et connecteurs
Disconnect and reconnect your flow path with ease.
  • Large ergonomic button and thumb pad with secure latch and optional integrated locking mechanism
  • Several connector configurations available in commonly used polypropylene and polycarbonate materials
  • Plugs and caps available to maintain fluid path integrity
RQ Series offers a cost effective solution for flexible tubing connections in single use systems. The connector set features a user-friendly interface that includes a large guarded button for a secure lock to guard against disconnects. Use these fittings to quickly connect and disconnect from your flow line. Simply order a body (female half) and an insert (male half).