Imprimer
Value Plastics® Certified Sanitary Adapters, ADCF Polypropylene
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX31513-18BLKEA 516 EUR
MFLX31513-18BLK MFLA31515-89BLK MFLA31515-90BLK MFLA31515-91BLK MFLA31515-92BLK MFLA31515-93BLK MFLA31515-95BLK MFLA31515-96BLK MFLA31515-97BLK MFLA31515-99BLK
Value Plastics® Certified Sanitary Adapters, ADCF Polypropylene
Raccords et connecteurs
Certificate of compliance included!
  • Animal derivative component free
  • Meet USP Class IV requirements
  • Sterilizable vis gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow—and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI.
Informations de livraison: Includes certificate of compliance.