Imprimer
Value Plastics® Adapter Fittings, Male Luer to Threaded, Straight
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX45518-82BLKEA 408 EUR
MFLX45518-82BLK MFLX45518-88BLK
Value Plastics® Adapter Fittings, Male Luer to Threaded, Straight
Raccords et connecteurs
Easily transition between luer fittings and UNF or NPT threads.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.