Imprimer
Halkey-Roberts® Thermoplastic Flow Ratchet Tubing Control Clamps
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX06833-01BLKEA 391 EUR
MFLX06833-01BLK MFLX06833-00BLK
Halkey-Roberts® Thermoplastic Flow Ratchet Tubing Control Clamps
Pinces Clips pour tubes
Won't rust, corrode, or distort.
  • Constructed of durable Celcon plastic
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.