Imprimer
Colour stripe silicone tubing, HelixMark®
Fournisseur: Freudenberg Medical Europe GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
61-079-21 61-079-27 61-080-21 61-080-23 61-080-26 61-080-27 61-080-30 61-080-32 61-079-23 61-079-26 61-079-30 61-079-32
HELI61-079-21EA 0 EUR
HELI61-079-21 HELI61-079-27 HELI61-080-21 HELI61-080-23 HELI61-080-26 HELI61-080-27 HELI61-080-30 HELI61-080-32 HELI61-079-23 HELI61-079-26 HELI61-079-30 HELI61-079-32
Colour stripe silicone tubing, HelixMark®
Tuyaux Transfer Tubing
Distinctive silicone colour striping enables users to recognise tube lines at a glance by application, tubing size, or functional area.
  • Fully autoclavable and sterilisable
  • Ideal for use in transfer operations as well as other applications requiring non-leachable material
  • Material certification in every package
  • Lot traceability
Designed to suit a wide array of laboratory applications or pharmaceutical processes. All tubes provide exceptional biocompatibility, having undergone studies according to ISO 10993-1 and meeting USP Class VI requirements. The production process ensures that the silicone stripe is integrated into the tube wall without touching or impeding the interior passage.
Certifications: Tubing is manufactured under strict good manufacturing practices (GMP) in a FDA registered medical device facility.
Informations de commande: Tubing is packaged in 15,24 m coils and double-bagged in crush-resistant boxes. Material certification and lot traceability is included in every box.