Imprimer
Silicone keyboards with integrated mouse button, InduProof® Advanced MB
Fournisseur: GETT GERATETECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
KG19259 KG21212
GETTKG19259EA 293 EUR
GETTKG19259 GETTKG21212
Silicone keyboards with integrated mouse button, InduProof® Advanced MB
Claviers
Rugged, waterproof and dustproof silicone keyboard with integrated mouse button.
  • Rugged, durable keyboard with 104 keys and integrated mouse button
  • Comfortable and ergonomic operation, even with protective gloves (silicone or latex)
  • IP 68 protection; device can be submerged into liquids
  • Easy cleaning and disinfection, even under tap water or by immersion
  • Increased chemical and mechanical resistance due to special surface treatment
  • VESA-M5-boreholes (distance 75×75mm)
  • 4-year warranty on technical function
  • Made in Germany
The InduProof® Advanced has a full set of keys with a numeric keyboard and an integrated mouse button (mini-joystick). It provides a pleasant sense of touch and a very high degree of protection. As a result of its robust design, it is highly suitable for use in challenging, hygiene sensitive areas like industrial food and drinks production. It is possible to completely clean and disinfect the unit. It has a long serviceable life as a result of its high-quality workmanship.
Certifications: CE, REACH, RoHs, EMC, IP 68.
Cdt: Keyboards are safely and environmentally friendly packed in brown cardboard boxes.