Imprimer
ESD cellulose tape, plastic core, Wescorp
Fournisseur: DESCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
242270 242271 242272 242275 242277
819-0143EA 9.6 EUR
819-0143 819-0144 819-0145 819-0146 819-0147
ESD cellulose tape, plastic core, Wescorp
Rubans adhésifs
Wescorp ESD cellulose tape, plastic core are used for general purpose ESD tape applications.
  • Prevents damage to sensitive electronic components in manufacturing
  • Secure (bundle) IC DIP tubes
  • Use with ESD symbols and/or yellow stripe for ESD awareness
  • Lead-free RoHS compliant
Ideal in packaging for container sealing, static shielding bag closure, and holding DIP tubes. Ideal for conformal coating or holding and sealing supplies in manufacturing.
Ideal for holding notes, work orders or instructions in offices, antistatic worksurfaces, or for general purpose third hand use.