Imprimer
Conductive Tote Boxes
Fournisseur: DESCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
239305 239345
DESC239305EA 26.7 EUR
DESC239305 DESC239345
Conductive Tote Boxes
Boîtes Boîtes antistatiques
Conductive tote boxes are made from injection moulded conductive polypropylene.
  • Flat base, stackable
  • Rv: 1×10³ to <1×10⁶ Ω
  • Custom-made dividers are available for PCB containers
  • Machined from approximately 2 mm thick conductive plastic or Cortronic® B-flute static dissipative cardboard
  • Many shapes and sizes possible
  • Process is suited for small and medium quantities