Imprimer
Metal-In shield bags, PCL100 Clean series
Fournisseur: Desco Industries
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DESCPCL1001115EA 46.7 EUR
DESCPCL1001115
Metal-In shield bags, PCL100 Clean series
Sachets Sacs de stockage
Metal-In static shielding bags are manufactured from a polyester, metal, polyethylene laminate. ESD safe packaging designed for use in cleanroom areas where surface particulate is of concern.
  • Embossed with ESD protective symbol and lot code information
  • The polyester dielectric in concert with the metal layer provides discharge shielding
  • Bag is free of amines, silicones and heavy metals
  • Bags are heat sealable
Metal-In film laminate 0,07 mm protects ESD sensitive contents from electrostatic fields and electrostatic discharges (ESD). Dissipative inner and outer surfaces allows bag to be used throughout an ESD Protected Area and grounded when placed on a grounded surface or handled by a grounded operator.
Certifications: Meets ANSI/ESD S20.20 and ANSI/ESD S541 requirements for ESD shielding packaging inside and outside an ESD Protected Area (EPA). Lead-free RoHS 3, REACH and Conflict Minerals Compliant.