Imprimer
Coil cord with 10 mm socket
Fournisseur: DESCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
230225 230245
115-3041EA 8.7 EUR
115-3041 115-3042
Coil cord with 10 mm socket
Dispositifs de protection antistatique Conducteurs de protection
Coil cord with 2,5 mm polyurethane insulation, conductor is high strength 7-end duplex tinsel for strength and flexibility.
  • 1 megohm resistor built in each socket (unless otherwise specified)
  • Dielectric strength: cords withstand 1250 volts DC for at least one minute
  • Colours: black or yellow