Imprimer
Flasks, flat bottom with Quickfit® ground joints
Fournisseur: BELLEVILLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
03372482 03372540 03372607
201-0309EA 68 EUR
201-0309 201-0310 201-0311
Flasks, flat bottom with Quickfit® ground joints
Ballons Ballons à fond plat
PYREX®, borosilicate glass, clear, with medium necks.
  • Very good chemical resistance
  • High temperature resistance
  • With longer neck than standard of flasks
Flat base allows use with hotplates. With labelling area.