Imprimer
Silicone encased keyboard for hygienic environments, InduProof® Smart Touch
Fournisseur: GETT GERATETECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
KG23221 KG24209 KG25200 KG25208
GETTKG23221EA 291 EUR
GETTKG23221 GETTKG24209 GETTKG25200 GETTKG25208
Silicone encased keyboard for hygienic environments, InduProof® Smart Touch
Claviers
Fully waterproof and dustproof medical keyboard with number block and touchpad.
  • Dustproof and waterproof keyboard with integrated capacitive touchpad
  • Can be completely disinfected
  • Soft typing through silicone switching elements
  • Flat key caps for thorough disinfection
  • Pleasant tactile feeling even with protective gloves
  • Three year warranty on technical function
  • Made in Germany
This InduProof® keyboard has a complete set of keys with numeric keypad, an integrated touchpad, and provides a pleasant tactile feel and the highest degree of protection. This makes the keyboard suitable for use in almost all hygiene-sensitive areas. It can be cleaned and disinfected all around. Thanks to the high quality ‘Made in Germany’ finish, it has a long service life. As part of the InduProof® brand family, it is manufactured in line with the highest quality standards. Customisation is also possible.
Certifications: CE, FCC, REACH, RoHs, EMC, IP 68.
Cdt: Keyboards are safely and environmentally friendly packed in brown cardboard boxes.