Imprimer
Silicone encased keyboards with full layout, integrated touch pad and backlit keys, GCQ CleanType® Prime Touch+
Fournisseur: GETT GERATETECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
KG26239 KG26236 KG26237 KG26238 KG26240 KG26241 KG26242 KG28220
GETTKG26239EA 195 EUR
GETTKG26239 GETTKG26236 GETTKG26237 GETTKG26238 GETTKG26240 GETTKG26241 GETTKG26242 GETTKG28220
Silicone encased keyboards with full layout, integrated touch pad and backlit keys, GCQ CleanType® Prime Touch+
Claviers
Silicone encased keyboard with full layout, integrated touch pad and backlit keys for use in highly hygienic sensitive areas. Completely waterproof and dust proof.
  • Integrated capacitive touch pad
  • Five-stage lighting function
  • Flexible mounting using magnets
  • EMC according to EN 60601-1-2 Edition 4
  • Keyboard meets IP 68 protection rating due to a fully enclosed silicone surface
  • Fully washable and disinfectable
  • Cleaning possible without unintentionally triggering keys by activating a keylock button
  • Two-year warranty
  • Engineered in Germany
The Cleantype® Prime Touch+ keyboard helps create a hygienic working environment. The surface is waterproof and dust proof and can therefore be thoroughly cleaned and disinfected.
This model keyboard is also equipped with special product features. It can be attached to magnetic surfaces in a flexible way and is suitable for darkened environments where space is cramped thanks to an incremental lighting function and the slim design. Thanks to the touch pad, there is no need to use an additional mouse. It provides the writing comfort of an office keyboard.
Certifications: CE, EMC, REACH, IP 68.
Informations de commande: Includes USB extension cable.
Cdt: Keyboards are packed in brown cardboard boxes.