Imprimer
Bags, zip seal, writing area, Minigrip®
Fournisseur: Flexico
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ITM020302 ITM020304 ITM020308 ITM020310 ITM020305 ITM020301 ITM020312 ITM020317 ITM020319 ITM020311 ITM020313 ITM020318 ITM020314 ITM020303 ITM020336 ITM020902 ITM020904 ITM020307 ITM020309
129-2009EA 97.5 EUR
129-2009 129-2010 129-2011 129-2012 129-2339 129-2016 129-2342 129-2345 129-2347 129-2341 129-2343 129-2346 129-2344 129-3861 129-3862 129-3863 129-3864 129-3865 129-3866
Bags, zip seal, writing area, Minigrip®
Sachets Sachets d'échantillonnage
Clear PE bags, thickness 60 µm, food contact.
  • High transparency and gloss
  • Suitable for food contact
  • 3 white write-on panels
  • Hanging and/or venting hole
  • Resealable, reusable, 100% recyclable
Made of polyethylene (LDPE). Zip seal designed for easy opening and high speed packing. The Minigrip® bag provides an excellent mechanical resistance and high temperature resistance (from –40 up to +60 °C).
Certifications: Suitable for food contact. Certificate available on request.