Imprimer
Pre-packed columns, 50 and 200 ml, SkillPak™
Fournisseur: TOSOH Bioscience
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0045300 0045301 0045302 0045303 0045304 0045305 0045306 0045307 0045308 0045309 0045310 0045311 0045312 0045313 0045314 0045315 0045316 0045317 0045318 0045319 0045320 0045321 0045322 0045323 0045324 0045325 0045326 0045327 0045328 0045329 0045330 0045331 0045332 0045333 0045334 0045335 0045336 0045337 0045338 0045339 0045340 0045341 0045342 0045343 0045344 0045345
TOYO0045300EA 3270 EUR
TOYO0045300 TOYO0045301 TOYO0045302 TOYO0045303 TOYO0045304 TOYO0045305 TOYO0045306 TOYO0045307 TOYO0045308 TOYO0045309 TOYO0045310 TOYO0045311 TOYO0045312 TOYO0045313 TOYO0045314 TOYO0045315 TOYO0045316 TOYO0045317 TOYO0045318 TOYO0045319 TOYO0045320 TOYO0045321 TOYO0045322 TOYO0045323 TOYO0045324 TOYO0045325 TOYO0045326 TOYO0045327 TOYO0045328 TOYO0045329 TOYO0045330 TOYO0045331 TOYO0045332 TOYO0045333 TOYO0045334 TOYO0045335 TOYO0045336 TOYO0045337 TOYO0045338 TOYO0045339 TOYO0045340 TOYO0045341 TOYO0045342 TOYO0045343 TOYO0045344 TOYO0045345
Pre-packed columns, 50 and 200 ml, SkillPak™
Colonnes pour chromatographie
SkillPak 50 and 200 ml columns are designed for fast method development and scale-up and are pre-packed with TOYOPEARL® and TSKgel® process chromatography media. These columns offer superior separation of biomelocules, such as monoclonal antibodies, proteins and oligonucleotides. They are the best solution to scale up methods developed on SkillPak 1 and 5 ml pre-packed columns.
  • Connections: 1/4-28 Standard fittings with flat-bottom geometry for 1/8” OD or 1/16” OD capillary
SkillPak pre-packed columns are designed for your purification and separation tasks from platform design to pilot scale. The columns are ready to use upon receipt and show excellent physical strength and ideal flow characteristics for industrial downstream process.
These columns guarantee optimal performance and can be operated with commonly used low or media pressure liquid chromatography systems. They are reproducibly packed and take into account the varying compressibility of each resin. This provides an accurate representation of conditions found in full scale columns.
Shipping buffer: 20 % ethanol for all TOYOPEARL and TSKgel resins.
Asymmetry factor (As): 0,8 to 1,4 for TOYOPEARL and TSKgel resins.