Imprimer
Spark® Cyto multimode plate readers with live cell imaging and real-time cytometry
Fournisseur: Tecan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
94000001 94100001 94200001 94300001 94400001
TCAT94000001EA 0 EUR
TCAT94000001 TCAT94100001 TCAT94200001 TCAT94300001 TCAT94400001
Spark® Cyto multimode plate readers with live cell imaging and real-time cytometry
Microplate Readers
Spark Cyto is the first multimode reader with live cell imaging and real time cytometry to offer as much information as possible from the same cells, under the same conditions.
  • Live cell imaging and real-time cytometry for 2D and 3D cell models
  • Temperature and gas control
  • Lid Lifter™
  • Enhanced Absorbance, Fluorescence and Luminescence modules
  • Fusion optics - combine the flexibility of monochromators with the sensitivity of filters
  • ALPHA technology
  • Fluorescence filter top/bottom
  • DNA-, RNA- & protein quantification
  • Low sample volume: 2 µl
  • Up to 16 sample in parallel
  • A260/280 purity check
  • Labeling effiency for nucleic acids
  • Full-spectra scan (Spark, SparkCyto)
Multimode plate reader with environmental control that makes multi-parametric measurement possible by combining standard detection modes (luminescence, fluorescence, absorbance) with imaging in 6- to 384- well formats.
Spark Cyto combines three magnification levels (2×, 4×, 10×) with four channels for fluorescence (blue, green, red and far-red) and bright field imaging, enabling high quality cell analysis for a wide range of applications.
Pre-defined imaging applications for the most common cytometric assays:
• Label-free cell counting
• 3Dai spheroid and organoid analysis
• Confluence
• Nuclei counting
• Transfection efficiency
• Cell viability
• Cell death
For kinetic measurements, Real-time Experimental Control (REC) automatically activates a procedure as soon as a specific data value is reached.
Environmental control: Temperature control (from RT to 42 °C), CO₂ and O₂ gas control
Imaging: Bright field, Digital phase contrast and Fluoresence imaging
Certifications: Carbon neutral certificate (ClimatePartner).
Informations de commande: PC, SparkControl™ and Image Analyser™ software included. Monitor, keyboard and mouse not included.
Informations de livraison: Supplied with a user manual, PC and power cable.
Attention: Spark Cyto is for Research Use Only. Not for use in diagnostic procedures.