Imprimer
Bags, zip seal, writing area, Minigrip® bio-based
Fournisseur: Flexico
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ITM010164 ITM010166 ITM010167 ITM010168 ITM010169 ITM010172 ITM010174 ITM010175 ITM010177
129-2280EA 72.1 EUR
129-2280 129-2281 129-2282 129-2283 129-2284 129-2285 129-2286 129-2287 129-2289
Bags, zip seal, writing area, Minigrip® bio-based
Sachets Sachets d'échantillonnage
Clear bio-based PE bags, thickness 50 or 60 µm, food safe.
  • Innovative, eco-designed and sustainable bag
  • Food safe
  • 3 white write-on panels
  • Hanging and/or venting hole
  • Resealable, reusable, 100% recyclable
Polyethylene LDPE from renewable natural resource (sugar cane). The bag is resealable, reusable and 100% recyclable (mono-material). The Minigrip® bio-based bag provides an excellent mechanical resistance and high temperature resistance (from –40 up to +60 °C).
Certifications: Food safe certificate available on request.