Imprimer
Quick-Disconnect Coupler Fittings, Hose Barbed, Straight, Dynalon®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX06290-05BLKEA 128 EUR
MFLX06290-05BLK MFLX06290-09BLK MFLX06290-10BLK
Quick-Disconnect Coupler Fittings, Hose Barbed, Straight, Dynalon®
Raccords et connecteurs
Provides easy single-handed disconnection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Construction from low-density polyethylene (LDPE).