Imprimer
Ergonomic computer mouse, GCQ® EasyMouse
Fournisseur: GETT GERATETECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
KH29210 KH29211
GETTKH29210EA 48.7 EUR
GETTKH29210 GETTKH29211
Ergonomic computer mouse, GCQ® EasyMouse
Souris d'ordinateur et tapis de souris
The EasyMouse is an ergonomic and reliable plastic mouse. It is equipped with two mouse buttons and a scroll wheel.
  • Medical grade IP 68 mouse
  • Completely waterproof and dustproof
  • Easy to clean and disinfect
  • Silicone cap protects USB connector from splashing water during cleaning
  • Ergonomic functionality with two mouse buttons and a scroll wheel
  • Symmetrical housing shape is suitable for right- and left-handed users
  • Optical sensor
  • Resolution: 1000 dpi
  • Button life >=2 million actuations
  • Engineered in Germany
The EasyMouse is an ergonomic and reliable plastic mouse. It is characterized by its good functionality and is equipped with two mouse buttons and a scroll wheel. Due to its symmetrical housing shape, it is equally well suited for right- and left-handed users. The EasyMouse is also completely waterproof and dustproof.
Certifications: CE, FCC, REACH, RoHs, IP68.
Cdt: The mouse is delivered safely packed in brown cardboard box.