Imprimer
PCR plates, 384-well, rigid frame, PC/PP, 0,03 ml, BIO-CERT® PCR QUALITY
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
781538 781539
732-3978EA 269 EUR
732-3978 732-3979
PCR plates, 384-well, rigid frame, PC/PP, 0,03 ml, BIO-CERT® PCR QUALITY
Microplaques Plaques PCR
These 384-well PCR plates are an economical solution for automatic high-throughput analysis.
  • High stability and rigidity of the plate due to the polycarbonate frame
  • Rapid heat transfer through thin vessel walls
  • For applications in automation and high-throughput analysis
The rigid frame PCR plates have a strong polycarbonate frame to ensure safe handling in semiautomated and automated pipetting systems and optimal evaporation protection. The rigid frame also allows the use of very thin-walled wells from 100% virgin medical grade polypropylene. The thin walls allow fast transmission of the temperature specified by the cycler, thus reducing cycle times.
For qPCR, white plates are available that optimally reflect the fluorescence signals. The cleanroom quality of the PCR consumables from BRAND guarantees reliable results. BRAND 384-well Rigid Frame plates are also produced under controlled cleanroom conditions and thus meet the purity criteria of the quality level BIO-CERT® PCR QUALITY.