Imprimer
Heating circulators, CORIO™ CP
Fournisseur: JULABO GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9013512 9013526 9013504 9013506
JULA9013512EA 3600 EUR
JULA9013512 JULA9013526 461-1458 JULA9013506
Heating circulators, CORIO™ CP
Thermostats à circulation Systèmes à circulation fermée Thermostats chauffants
The powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision, and a wide working temperature range. The CORIO™ CP model is an advanced unit with professional technology for demanding applications and can be used for temperature control of internal or external applications due to the powerful and infinitely adjustable pressure pump. The jet nozzle allows continuous adjustment of the pump stream in the system. These heating circulators are optimised for external temperature applications in combination with jacketed reactors, distillation apparatus, mini-plant applications, photometers, refractometers and can also be used for internal temperature applications of samples and small objects.
  • Bright, white, easy to read display
  • Very quiet
  • Easy switching between internal and external circulation
  • USB connection
  • RS232 interface for online communication
  • High-quality bath tanks made of stainless steel with bath lid and drain tap
  • Integrated cooling coil for counter-cooling
Certifications: Classification III according to DIN 12876-1
Informations sur les accessoires: An extensive range of accessories mean that the circulators can be modularly and individually adapted to applications.
Informations de livraison: Supplied with two barbed fittings for tubing 8 and 12 mm Int.Ø. (Pump connections M16×1 male).