Imprimer
Disposable socks, Foot Fresh, Hygostar
Fournisseur: MENSCH FRANZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
28700 28701
MENS28700EA 0 EUR
MENS28700 114-9381
Disposable socks, Foot Fresh, Hygostar
Vêtements à usage unique
Disposable socks made of 100% polyamide. Can be used as surgical socks.
  • Hygienic and breathable
  • Very stretchy, excellent fit
  • Latex-free
  • Retain their shape, comfortable to wear for several hours
  • Polyamide is resistant to bacteria and fungi
The disposable socks are perfect for patients as well as hygienic surgical socks for nursing staff to wear in their professional shoes. They are for single use and are disposed of at the end of the working day, so germs and contamination don't stand a chance.
Cdt: One pair per bag.