Imprimer
Advanced polyethylene CentriStar™ cap for centrifuge tubes, Corning®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
431471 431473
525-0478EA 68.1 EUR
525-0478 525-0480
Advanced polyethylene CentriStar™ cap for centrifuge tubes, Corning®
Capuchons Bouchons pour tubes à essais
The CentriStar™ Cap has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping.
This leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when used under recommended conditions.