Imprimer
J.T.Baker®, Robotic Tips, Type HT 2
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
49008-0015EA 423 EUR
49008-0015 49008-0115 49008-1015 49008-1115 49009-0015 49009-0115 49009-1015 49009-1115 49010-0015 49010-0115 49010-1015 49010-1115
J.T.Baker®, Robotic Tips, Type HT 2
Pointes de pipettes
Suitable for use on third party robotic instruments Hamilton Microlab Star and Nimbus. Available in conductive version, with or without filter.
  • Manufactured under cleanroom conditions (cleanroom Class 8)
  • State-of-the-art quality control
  • Ideal for high throughput analysis
  • Barcoded trays for automated identification
  • Packed in blister
J.T.Baker® robotic tips type HT 2 are suitable for third party robotic instruments Hamilton Microlab Star, Nimbus, and other robotic instruments using Hamilton’s CO-RE or CO-RE II pipetting head.
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Hamilton, Microlab Star™, Nimbus® and CO-RE® are registered trademarks of Hamilton company.