Imprimer
Deep well and assay plates, Axygen®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P-2ML-SQ-C P-2ML-SQ-C-S P-384-240SQC P-384240SQCS P-5ML-48-C P-5ML-48-C-S P-96-450R-C P-96-450R-C-S P-96-450V-C P-96-450V-C-S P-DW-11-C-S P-DW-20CSIND P-DW-500-C P-384-120SQ-C P-DW-11-C P-DW-20-C P-DW-20-C-S P-DW-12-HC P-DW-12-HC-S
736-0344EA 171 EUR
736-0344 736-0345 736-0332 738-1000 736-0348 736-0349 732-1370 732-1371 732-1372 732-1373 736-0339 738-0187 736-0335 736-0357 736-0358 736-0359 737-0159 738-0185 738-0186
Deep well and assay plates, Axygen®
Microplaques Microplaques deepwell
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
  • 0,6 ml deep well plate
  • ANSI/SLAS 1-2004: Microplates - footprint dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.