Imprimer
Garment cupboards
Fournisseur: TEKNOMEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SHCBGRS106017 SHCBGRS106020 SHCBGRS126017 SHCBGRS126020 SHCBGSS106017 SHCBGSS106020 SHCBGSS126017 SHCBGSS126020
TEKMSHCBGRS106017EA 0 EUR
TEKMSHCBGRS106017 TEKMSHCBGRS106020 TEKMSHCBGRS126017 TEKMSHCBGRS126020 TEKMSHCBGSS106017 TEKMSHCBGSS106020 TEKMSHCBGSS126017 TEKMSHCBGSS126020
Garment cupboards
Mobilier Armoires de stockage
Our garment cupboard comes complete with hanging rail.
  • Sloping top
  • Brushed dull polish finish
  • Stainless steel adjustable feet with non-marking rubber base shelving optional for half rail
Our garment cupboard with solid double doors is ideal to store clothes securely in environments that require robust, hygienic and easy-clean stainless steel furniture.
It comes with a hanging rail (full or half rail) and is manufactured from 1,5 mm thick stainless steel, set in a super strong frame making these cupboards incredibly robust. And for the half rail you have the option to add shelving if required.
Moreover, they are designed with hygiene in mind too. No ledges or crevices assures that dirt cannot collect. The solid stainless steel construction and sloping top enable easy cleaning and the 300 mm legs allow easy access for cleaning underneath.