Imprimer
Advanced TOUCH, InduProof®
Fournisseur: GETT GERATETECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
KG17201 KG17202
GETTKG17201EA 318 EUR
GETTKG17201 GETTKG17202
Advanced TOUCH, InduProof®
Claviers
Rugged, water- and dust-proof silicone keyboard with integrated touchpad.
  • Rugged, durable keyboard with 104 keys and integrated mouse button
  • Comfortable and ergonomic operation also with protective gloves made of silicone or latex
  • IP 68 protection, can be submerged into liquids
  • Easy cleaning and disinfection - even under tap water or by immersion
  • Increased chemical & mechanical resistance due to special surface treatment
  • VESA-M5-boreholes (distance 75×75 mm)
  • 4 years warranty on technical function
  • Made in Germany
The InduProof® Advanced keyboard has a complete set of keys with numeric keypad, an integrated touchpad and convinces with a pleasant feel and the highest degree of protection. Its robust design makes it ideal for use in demanding hygiene-sensitive areas such as industrial food and beverage production. High-quality workmanship and the use of high-quality materials in this keyboard ensure a long service life.
Certifications: CE, REACH, RoHs, EMC, IP 68.
Cdt: The keyboards are packed in brown cardboard boxes.