Imprimer
Collagen I-coated microplates, Corning® BioCoat®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
354664 354665 354666 354667 356407 356519 356705 356702 356700 356699 356698 356703 354407 354519 354649 354650 356649 356650 356664 356665 356666 356667
734-0261EA 284 EUR
734-0261 734-0262 734-0263 734-0264 734-0276 734-0303 734-1136 734-1133 734-1131 734-1130 734-1129 734-1134 734-0114 734-0179 734-0248 734-0249 734-0319 734-0320 734-0329 734-0330 734-0331 734-0332
Collagen I-coated microplates, Corning® BioCoat®
Microplaques Microplaques pour culture cellulaire
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture
  • Storage: 4 to 30 °C under dry conditions