Imprimer
Clean sterile disposable boot covers, CleanMax®
Fournisseur: LAKELAND CLEANMAX APPAREL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CTL903CSP-2X CTL903CSP-L/X CTL903CSP-S/M
114-9548EA 525 EUR
114-9548 114-9549 114-9550
Clean sterile disposable boot covers, CleanMax®
Vêtements à usage unique
Clean and Sterile disposable boot covers are individually double-bagged in pairs and folded to promote easier gowning with less excursions.
  • Clean and sterile in silicone-free environment
  • Latex-free
  • Tunnelled elastic top
  • 19" High
  • Non-skid PVC sole
  • Individually double-bagged in pairs
  • Gamma radiation indicator dots on each package
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparel provides chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certifications: Compatible for ISO Class 5 – 8 cleanrooms and all controlled environments.