Imprimer
HyFlex® 11-280 Cut Resistant Sleeves without Thumbslot, INTERCEPT™
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11-280/12N 11-280/12W 11-280/16N 11-280/16W 11-280/18N 11-280/18W
ANSE11-280/12NEA 533 EUR
ANSE11-280/12N ANSE11-280/12W ANSE11-280/16N ANSE11-280/16W ANSE11-280/18N ANSE11-280/18W
HyFlex® 11-280 Cut Resistant Sleeves without Thumbslot, INTERCEPT™
Tenue de travail
High cut protection sleeve with excellent breathability.
  • INTERCEPT™ Cut resistance technology provides best-in-class cut protection and a soft and cool feel to the worker
  • Seamless design and the options of lengths and widths ensure an excellent fit
  • Sleeves are engineered to perform in combination with any HyFlex® cut resistant glove
  • Available in wide or narrow versions
  • Compliant with FDA food handling regulations 21 CFR 177.1520 and 21 CFR 177.2600
INTERCEPT™ cut resistance technology has been tested to perform at or exceed industry standards. This Ansell proprietary technology offers best-in-class cut protection and increased comfort. The seamless design and the width options make these sleeves an ideal choice for all-day comfort.