Imprimer
Standard coveralls with collar, BioClean-D™
Fournisseur: Nitritex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S-BDCCT-3XL S-BDCCT-4XL S-BDCCT-5XL S-BDCCT-6XL S-BDCCT-7XL
NITRS-BDCCT-3XLEA 358 EUR
NITRS-BDCCT-3XL NITRS-BDCCT-4XL NITRS-BDCCT-5XL NITRS-BDCCT-6XL NITRS-BDCCT-7XL
Standard coveralls with collar, BioClean-D™
Vêtements à usage unique
Sterile, disposable coverall with collar for protection against chemicals.
  • Anti-static, lightweight, low-linting Cleantough material
  • Thumb loops to ensure a secure hold
  • Zip with sealable cover
  • Elasticated back, cuffs, and ankles
The BioClean-D Coverall with collar features a front zip with protective flap, elasticated back, cuffs and ankles, and thumb loops to ensure a secure hold. The anti-static, lightweight, low-linting CleanTough material provides comfort and protection from a range of chemicals.