Imprimer
Cut resistant sleeve with thumbslot, INTERCEPT™, HyFlex® 11-281 series
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11-281/12N 11-281/12W 11-281/16N 11-281/16W 11-281/18N 11-281/18W
ANSE11-281/12NEA 534 EUR
ANSE11-281/12N ANSE11-281/12W ANSE11-281/16N ANSE11-281/16W ANSE11-281/18N ANSE11-281/18W
Cut resistant sleeve with thumbslot, INTERCEPT™, HyFlex® 11-281 series
Tenue de travail
The most comfortable sleeve with INTERCEPT™ technology.
  • INTERCEPT™ Cut resistance technology provides best-in-class cut protection and a soft and cool feel to the worker
  • Seamless design and the options of lengths and widths ensure an excellent fit
  • Sleeves are engineered to perform in combination with any HyFlex® cut resistant glove
  • Available in wide or narrow versions
INTERCEPT™ Cut resistance technology has been tested to perform at or exceed industry standards. This Ansell proprietary technology offers best-in-class cut protection and increased comfort. The seamless design and the width options make these sleeves an ideal choice for all-day comfort.
Certifications: Compliant with FDA food handling regulations 21 CFR 177.1520 and 21 CFR 177.2600.