Imprimer
Industrial natural rubber latex gloves, AlphaTec® 87-086 series
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
87-086/6.5 87-086/7.5 87-086/8.5
ANSE87-086/6.5EA 36.7 EUR
ANSE87-086/6.5 ANSE87-086/7.5 ANSE87-086/8.5
Industrial natural rubber latex gloves, AlphaTec® 87-086 series
Gants
Lightweight natural rubber gloves offering comfort and food compliance.
  • Beaded cuff for tear resistance and easy donning
  • Food contact approved
  • Good abrasion resistance
  • 100% cotton flock lining
  • AQL 0.65
Flock lined, lightweight natural rubber latex gloves are designed for use in low hazard areas. Resistant, flexible and very comfortable.