Imprimer
Chemical and heat resistant gloves, AlphaTec® 37-340
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
37-340/7.5 37-340/10.5
ANSE37-340/7.5EA 0 EUR
ANSE37-340/7.5 ANSE37-340/10.5
Chemical and heat resistant gloves, AlphaTec® 37-340
Gants
Excellent resistance to many solvents and oil-based chemicals.
  • Very good abrasion resistance
  • Allows contact up to 100 °C
  • Superior raised grip pattern for safety handlings
  • Open cuff with textile edge binding allows ventilation to keep hand cool
These supported chemical gloves offer very good abrasion resistance. They feature an open cuff with textile edge binding to allow ventilation, keeping hands cool. Gloves allow contact up to 100 °C, and have a superior raised grip pattern for safe handling of materials.