Imprimer
ISO AIR cleanroom masks with duckbill
Fournisseur: Medicom
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60.754L 60.754M
114-9496EA 223 EUR
114-9496 114-9495
ISO AIR cleanroom masks with duckbill
Masques Masques pour salles propres
ISO AIR cleanroom masks with duckbill feature a large and comfortable breathing chamber.
  • Made in France
  • Made from 30% recycled polypropylene material
  • High breathability
  • ISO 3 Grade B protection
Masks feature low particle release, and are Helmke drum tested 'excellent' for filtration efficiency (BFE, PFE, VFE >99%). Softex technology limits the risk of particulate contamination while reducing the risk of skin irritation. An adjustable and malleable nose clip prevents glasses from fogging up. Duckbill-style masks feature a large and comfortable breathing chamber, for increased wearer comfort over extended use.
Certifications: The ISO AIR range is manufactured in an ISO 8 cleanroom.
Cdt: 6 double packed bags × 50 units.