Imprimer
Self-standing centrifuge tubes, 50 ml, Corning®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
430897 430921
734-1872EA 223 EUR
734-1872 734-1876
Self-standing centrifuge tubes, 50 ml, Corning®
Tubes Tubes à centrifuger
Corning® 50 ml centrifuge tubes feature black printed graduations and a large white marking spot.
  • Skirted for self-standing
  • Disposable, sterile
  • Certified non pyrogenic
  • RNase- and DNase-free
Choose from plug-seal cap (95 kPa pressure-tested) or easy on/off CentriStar™ flat-top caps.
They have a conical bottom for pelleting and sample recovery.